• USA Home
  • 324795 - N-Hydroxysuccinimidyl acetoacetate

EMAIL THIS PAGE TO A FRIEND
324795 Aldrich

N-Hydroxysuccinimidyl acetoacetate

98%

Purchase

Properties

Related Categories Active Esters/Additives, Chemical Biology, Chemical Synthesis, Coupling, Peptide Chemistry,
InChI Key   VOIVNIGJPPFYCC-UHFFFAOYSA-N
assay   98%
mp   122-124 °C(lit.)
storage temp.   2-8°C

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?