• USA Home
  • 328073 - Cellulose acetate phthalate

EMAIL THIS PAGE TO A FRIEND
328073 Aldrich

Cellulose acetate phthalate

  • CAS Number 9004-38-0

  • Empirical Formula (Hill Notation) C116H116O64

  • Molecular Weight 2534.12

  •  MDL number MFCD00072769

  • Popular Documents:  FTIR (PDF)  

Purchase

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

22192 Cellulose acetate phthalate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?