• USA Home
  • 328898 - 9-Fluoreneacetic acid

EMAIL THIS PAGE TO A FRIEND
328898 Aldrich

9-Fluoreneacetic acid

99%

Synonym: 9-Fluorenylacetic acid

Purchase

Properties

Related Categories Building Blocks, C13 to C42+, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   WFSMJMTYIMFHPV-UHFFFAOYSA-N
assay   99%
mp   133-135 °C(lit.)
Gene Information   rat ... Akr1b1(24192)

Description

Packaging

1 g in glass bottle

Application

9-Fluoreneacetic acid was used in preparation of pyrokinin class of insect neuropeptides. It was also used in preparation of N-(9-fluoreneacetyl)-L-leucine.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?