• USA Home
  • 329223 - 2′-Amino-4′,5′-dimethoxyacetophenone

EMAIL THIS PAGE TO A FRIEND
329223 Aldrich

2′-Amino-4′,5′-dimethoxyacetophenone

99%

  • CAS Number 4101-30-8

  • Empirical Formula (Hill Notation) C10H13NO3

  • Molecular Weight 195.22

  •  PubChem Substance ID 329754389

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   KGKWXEGYKGTMAK-UHFFFAOYSA-N
assay   99%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?