• USA Home
  • 329312 - 3,5-Dimethylbenzenethiol

EMAIL THIS PAGE TO A FRIEND
329312 Aldrich

3,5-Dimethylbenzenethiol

96%

Synonym: 3,5-Dimethylthiophenol

Purchase

Properties

InChI Key   CESBAYSBPMVAEI-UHFFFAOYSA-N
assay   96%
refractive index   n20/D 1.568(lit.)
bp   127.5 °C/50 mmHg(lit.)
density   1.015 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
185 °F
Flash Point(C) 
85 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?