Synonym: Pd/C
Linear Formula Pd
Molecular Weight 106.42
MDL number MFCD03457879
PubChem Substance ID 24859834
Popular Documents: Specification Sheet (PDF)
| Related Categories | Catalysis and Inorganic Chemistry, Chemical Synthesis, Heterogeneous Pd Catalysts, Palladium |
| contains | ~50% water |
| extent of labeling | 5 wt. % loading (dry basis) |
| weight | Unit weight excludes weight of water |
| matrix | activated carbon, wet support |
Reagent especially useful in the reduction of fatty acids and bulky molecules.
catalyst for hydrogenation and hydrogenolysis
Catalyst for:
• Stille reaction1
• Hydrogenation reactions2
• Suzuki-Miyaura cross-coupling and Heck-Mizoroki reactions3
• Deoxygenation4
• Oxidation reactions5
• Coupling reactions6
10, 50 g in glass bottle
Customers Also Viewed

extent of labeling: 10 wt. % loading (dry basis), matrix activated carbon, wet support, Degussa type E101 NE/W

extent of labeling: 5 wt. % loading (dry basis), matrix activated carbon, wet support

extent of labeling: 5 wt. % loading (dry basis), matrix activated carbon, wet support

extent of labeling: 5 wt. % loading (dry basis), matrix activated carbon, wet support, Degussa type E101 NO/W
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?