EMAIL THIS PAGE TO A FRIEND
331481 Aldrich

Anthracene

zone-refined, ≥99%

Synonym: Anthraxcene, Paranaphthalene

  • CAS Number 120-12-7

  • Empirical Formula (Hill Notation) C14H10

  • Molecular Weight 178.23

  •  Beilstein Registry Number 1905429

  •  Colour Index Number 10790

  •  EC Number 204-371-1

  •  MDL number MFCD00001240

  •  PubChem Substance ID 24859965

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Arenes, Building Blocks, Chemical Synthesis, Light Emitters and Dopants, Materials Science,
grade   zone-refined
vapor density   6.15 (vs air)
vapor pressure   1 mmHg ( 145 °C)
InChI Key   MWPLVEDNUUSJAV-UHFFFAOYSA-N
assay   ≥99%
form   crystalline
autoignition temp.   1004 °F
bp   340 °C(lit.)
mp   210-215 °C(lit.)
solubility   alcohols: soluble
  benzene: soluble
  chloroform: soluble
  hydronaphthalenes: soluble
  supercritical carbon dioxide: soluble
Orbital energy   HOMO 5.7 eV 
  LUMO 1.7 eV 
Gene Information   human ... CYP1A2(1544)

Description

Packaging

250 mg in glass bottle

Application

Anthracene has been shown to be soluble in a variety of binary and ternary mixtures of cyclohexanone, ethyl acetate, and methanol .

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
CA9350000
Flash Point(F) 
249.8 °F
Flash Point(C) 
121 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

comparable product

Product #

Description

Add to Cart

694959 Anthracene, sublimed grade, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?