• USA Home
  • 331554 - Boron trifluoride methanol-d4 complex

EMAIL THIS PAGE TO A FRIEND
331554 Aldrich

Boron trifluoride methanol-d4 complex

in excess methanol-d4, ≥99 atom % D

Purchase

Properties

Related Categories Acids, Acids & Bases, Boron, Catalysis and Inorganic Chemistry, Chemical Synthesis,
InChI Key   JBXYCUKPDAAYAS-LHHVLQQYSA-N
isotopic purity   ≥99 atom % D
composition   BF3, 12 wt. %
concentration   in excess methanol-d4
refractive index   n20/D 1.326(lit.)
density   0.936 g/mL at 25 °C(lit.)
mass shift   M+4

Description

Application

Reagent for preparing deuterated methyl esters.

Packaging

5 g in ampule

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Price and Availability

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service.
Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 3286 8(6.1)(3) / PGII
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?