• USA Home
  • 331694 - Dibromobis(tributylphosphine)nickel(II)

EMAIL THIS PAGE TO A FRIEND
331694 Aldrich

Dibromobis(tributylphosphine)nickel(II)

97%

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Nickel More...
InChI Key   JRHYDBOXJQOUJY-UHFFFAOYSA-L
assay   97%
mp   55-56 °C(lit.)

Description

Packaging

25 g in glass bottle

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?