332445 Aldrich

Boron

amorphous, powder, (submicron)

DOWNLOAD MSDS (PDF)
  • CAS Number 7440-42-8

  • Empirical Formula (Hill Notation) B

  • Molecular Weight 10.81

  •  EC Number 231-151-2

  •  MDL number MFCD00134034

  •  PubChem Substance ID 24859897

Properties

Related Categories Electronic Chemicals, Materials Science, Micro/NanoElectronics, Pure Elements
form   amorphous
  powder
resistivity   1.5E12 μΩ-cm20 °C
particle size   (submicron)
density   2.34 g/mL at 25 °C(lit.)

Description

Packaging

5 g in glass bottle

Price and Availability

Customers Also Viewed

Boron
266620Boron

crystalline, 1 cm, 99.7% trace metals basis

Boron
333360Boron

crystalline, −60 mesh, 99% trace metals basis

Boron
15566Boron

purum p.a., ≥99.0%, pieces, crystalline

Boron
15580Boron

≥95% (boron), amorphous powder

Boron carbide

powder, −200 mesh, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?