• USA Home
  • 332534 - Tetrafluorophthalic acid

EMAIL THIS PAGE TO A FRIEND
332534 Aldrich

Tetrafluorophthalic acid

90%

Purchase

Properties

Related Categories Building Blocks, C8, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   YJLVXRPNNDKMMO-UHFFFAOYSA-N
assay   90%
impurities   5-10% water
mp   152-154 °C(lit.)

Description

Packaging

5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

196800 Tetrafluorophthalic acid, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?