• USA Home
  • 333042 - 2,5-Bis(trifluoromethyl)benzoyl chloride

EMAIL THIS PAGE TO A FRIEND
333042 Aldrich

2,5-Bis(trifluoromethyl)benzoyl chloride

97%

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
InChI Key   LRJNPOCQOZWIGR-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.4315(lit.)
density   1.528 g/mL at 25 °C(lit.)

Description

Packaging

5 g in glass bottle

Application

2,5-Bis(trifluoromethyl)benzoyl chloride may be used in chemical synthesis.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
Flash Point(F) 
165.2 °F
Flash Point(C) 
74 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?