• USA Home
  • 334189 - 3-Amino-2-methylbenzoic acid

EMAIL THIS PAGE TO A FRIEND
334189 Aldrich

3-Amino-2-methylbenzoic acid

97%

Synonym: 3-Amino-o-toluic acid

  • CAS Number 52130-17-3

  • Linear Formula H2NC6H3(CH3)CO2H

  • Molecular Weight 151.16

  •  Beilstein Registry Number 2803090

  •  MDL number MFCD00075026

  •  PubChem Substance ID 24860061

  •  eCl@ss 32160406

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Aromatic Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   BYHMLZGICSEKIY-UHFFFAOYSA-N
assay   97%
mp   178-181 °C(lit.)

Description

Packaging

50 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?