• USA Home
  • 334952 - (Bicyclo[2.2.1]hepta-2,5-diene)[(2S,3S)-bis(diphenylphosphino)butane]rhodium(I) perchlorate

EMAIL THIS PAGE TO A FRIEND
334952 Aldrich

(Bicyclo[2.2.1]hepta-2,5-diene)[(2S,3S)-bis(diphenylphosphino)butane]rhodium(I) perchlorate

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Hydrogenation
InChI Key   WGNSDFDYTHJVJG-HFYJLKHYSA-M
mp   210 °C (dec.)(lit.)

Description

Packaging

25, 100 mg in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1479 5.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?