• USA Home
  • 335053 - Ferroceneacetonitrile

EMAIL THIS PAGE TO A FRIEND
335053 Aldrich

Ferroceneacetonitrile

Synonym: Ferrocenylmethyl cyanide, NSC 301053

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Iron More...
InChI Key   UBCZLWARZCERJD-UHFFFAOYSA-N
mp   81-83 °C(lit.)

Description

Packaging

1 g in glass bottle

Application

Uses:
• Preparation of redox-active bis(ferrocenyl-amidine) anion sensor
• Electrochemical detection of nucleic acids (DNA sensor)
• Reactant in electron-transfer reactions
• Reactant in electrooxidation reactions (indirect oxidation of the side chain)

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?