EMAIL THIS PAGE TO A FRIEND
335118 Aldrich

Furfuryl benzoate

97%

Synonym: 2-Furylmethyl benzoate

Purchase

Properties

Related Categories Building Blocks, C12 to C20+, Chemical Synthesis, Furans, Heterocyclic Building Blocks More...
InChI Key   OYUZXJBNSJHTRF-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.544(lit.)
bp   117-119 °C/1.5 mmHg(lit.)
density   1.177 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?