EMAIL THIS PAGE TO A FRIEND
33527 Aldrich

Dibenzo-15-crown-5

≥97.0% (GC)

  • CAS Number 14262-60-3

  • Empirical Formula (Hill Notation) C18H20O5

  • Molecular Weight 316.35

  •  Beilstein Registry Number 1690434

  •  MDL number MFCD00055949

Purchase

Properties

Related Categories Chelation/Complexation Compounds, Chemical Synthesis, Crown Ethers, Synthetic Reagents
InChI Key   QSBFECWPKSRWNM-UHFFFAOYSA-N
assay   ≥97.0% (GC)

Description

Packaging

1 g in glass bottle

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Cross-Coupling Guide
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?