• USA Home
  • 337412 - (R)-(−)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl p-toluenesulfonate

EMAIL THIS PAGE TO A FRIEND
337412 Aldrich

(R)-(−)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl p-toluenesulfonate

97%

Synonym: (R)-2,2-Dimethyl-1,3-dioxolane-4-methyl p-toluenesulfonate, 1,2-Isopropylidene-sn-glycerol 3-tosylate, D-α,β-Isopropylideneglycerol γ-tosylate

  • CAS Number 23788-74-1

  • Empirical Formula (Hill Notation) C13H18O5S

  • Molecular Weight 286.34

  •  Beilstein Registry Number 89800

  •  MDL number MFCD00063233

  •  PubChem Substance ID 24860479

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Protected Alcohols More...
InChI Key   SRKDUHUULIWXFT-LLVKDONJSA-N
assay   97%
optical activity   [α]20/D −7.0°, neat
refractive index   n20/D 1.506(lit.)
mp   41-43 °C(lit.)
density   1.208 g/mL at 25 °C(lit.)

Description

Packaging

250 mg in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

59453 (R)-2,2-Dimethyl-1,3-dioxolane-4-methanol p-toluenesulfonate, ≥99.0% (sum of enantiomers, GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?