• USA Home
  • 337420 - (S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl p-toluenesulfonate

EMAIL THIS PAGE TO A FRIEND
337420 Aldrich

(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl p-toluenesulfonate

97%

Synonym: (S)-2,2-Dimethyl-1,3-dioxolane-4-methyl p-toluenesulfonate, 2,3-Isopropylidene-sn-glycerol 1-tosylate, L-α,β-Isopropylideneglycerol-γ-tosylate

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Protected Alcohols More...
InChI Key   SRKDUHUULIWXFT-NSHDSACASA-N
assay   97%
optical activity   [α]26/D +7.5°, c = 2% in DMF
refractive index   n20/D 1.506(lit.)
density   1.208 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Packaging

1, 5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?