• USA Home
  • 338796 - Ammonium phosphate dibasic

EMAIL THIS PAGE TO A FRIEND
338796 Aldrich

Ammonium phosphate dibasic

99%

Synonym: Ammonium hydrogenphosphate, Diammonium hydrogenphosphate, di-Ammonium hydrogenphosphate (sec)

Purchase

Properties

InChI Key   MNNHAPBLZZVQHP-UHFFFAOYSA-N
assay   99%
mp   155 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?