• USA Home
  • 339849 - Sulfur dichloride solution

EMAIL THIS PAGE TO A FRIEND
339849 Aldrich

Sulfur dichloride solution

80%, technical grade, 1.0 M in methylene chloride

Purchase

Properties

grade   technical grade
vapor density   3.55 (vs air)
vapor pressure   25.67 psi ( 55 °C)
  7.64 psi ( 20 °C)
InChI Key   FWMUJAIKEJWSSY-UHFFFAOYSA-N
assay   80%
concentration   1.0 M in methylene chloride
density   1.355 g/mL at 25 °C

Description

Packaging

Packaged under nitrogen in Sure/Seal bottles

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3390 6.1/PG 1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?