EMAIL THIS PAGE TO A FRIEND
340197 Aldrich

Inhibitor remover

Purchase

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

WGK Germany 
nwg
RTECS 
BD1200000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?