• USA Home
  • 343544 - (R)-(−)-Dihydro-5-(hydroxymethyl)-2(3H)-furanone

EMAIL THIS PAGE TO A FRIEND
343544 Aldrich

(R)-(−)-Dihydro-5-(hydroxymethyl)-2(3H)-furanone

97%

Synonym: (R)-γ-Hydroxymethyl-γ-butyrolactone, (R)-4,5-Dihydro-5-hydroxymethyl-2(3H)-furanone, (R)-5-Hydroxymethyl-2-oxotetrahydrofuran

  • CAS Number 52813-63-5

  • Empirical Formula (Hill Notation) C5H8O3

  • Molecular Weight 116.12

  •  Beilstein Registry Number 1680285

  •  MDL number MFCD00040528

  •  PubChem Substance ID 24861173

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Furans, Heterocyclic Building Blocks More...
InChI Key   NSISJFFVIMQBRN-SCSAIBSYSA-N
assay   97%
optical activity   [α]20/D −56°, c = 3 in chloroform
refractive index   n20/D 1.47(lit.)
bp   101-102 °C/0.048 mmHg(lit.)
density   1.237 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Packaging

1 g in glass bottle

Application

Versatile chiral synthon used in the synthesis of natural and unnatural products.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?