EMAIL THIS PAGE TO A FRIEND
345148 Aldrich

Nessler’s reagent

Synonym: Dipotassium tetraiodomercurate(II) in dilute sodium hydroxide

  • CAS Number 7783-33-7

  • Empirical Formula (Hill Notation) K2HgI4

  • Molecular Weight 786.40

  •  MDL number MFCD00049656

  •  PubChem Substance ID 24861297

Purchase

Properties

InChI Key   OPCMAZHMYZRPID-UHFFFAOYSA-J
density   1.097 g/mL at 25 °C(lit.)

Description

Application

For ammonium analysis. Allow to settle and use the clear supernatant liquid

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN2922 - class 8 - PG 2 - EHS - basic - Corrosive liquids, toxic, n.o.s., HI: all
WGK Germany 
2
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

72190 Nessler’s reagent, for qualitative det. of ammonia and ammonium compounds

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?