• USA Home
  • 345997 - 4,5-Dimethoxy-2-nitrocinnamic acid

EMAIL THIS PAGE TO A FRIEND
345997 Aldrich

4,5-Dimethoxy-2-nitrocinnamic acid

97%

Purchase

Properties

Related Categories Building Blocks, C11 to C12, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   BZIRMMAJZSOLEW-ONEGZZNKSA-N
assay   97%
mp   293 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?