EMAIL THIS PAGE TO A FRIEND
346209 Aldrich

1-Octadecanol

technical grade, 80%

Synonym: Octadecyl alcohol, Stearyl alcohol

Purchase

Properties

grade   technical grade
vapor density   9.3 (vs air)
vapor pressure   <0.01 mmHg ( 38 °C)
InChI Key   GLDOVTGHNKAZLK-UHFFFAOYSA-N
assay   80%
autoignition temp.   842 °F
expl. lim.   ~8 %
mp   56-59 °C(lit.)
density   0.812 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
RG2010000
Flash Point(F) 
383 °F
Flash Point(C) 
195 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?