• USA Home
  • 346330 - trans-3-Bromo-N-ethylcinnamamide

EMAIL THIS PAGE TO A FRIEND
346330 Aldrich

trans-3-Bromo-N-ethylcinnamamide

99%

Purchase

Properties

Related Categories Amides, Building Blocks, C8 to C20+, Carbonyl Compounds, Chemical Synthesis,
InChI Key   LDCXGZCEMNMWIL-VOTSOKGWSA-N
assay   99%
mp   89-91 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
2
RTECS 
UC6314000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?