• USA Home
  • 346365 - Boc-D-Met-OH (dicyclohexylammonium) salt

EMAIL THIS PAGE TO A FRIEND
346365 Aldrich

Boc-D-Met-OH (dicyclohexylammonium) salt

98%

Synonym: N-(tert-Butoxycarbonyl)-D-methionine (dicyclohexylammonium) salt, Boc-D-methionine (dicyclohexylammonium) salt

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Methionine, Peptide Chemistry More...
InChI Key   SKDIPRMGDVUUQO-HMZWWLAASA-N
assay   98%
optical activity   [α]20/D −27°, c = 1 in chloroform
mp   141-143 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?