• USA Home
  • 348368 - (S)-(−)-1-(2-Methoxybenzoyl)-2-(methoxymethyl)pyrrolidine

EMAIL THIS PAGE TO A FRIEND
348368 Aldrich

(S)-(−)-1-(2-Methoxybenzoyl)-2-(methoxymethyl)pyrrolidine

99%

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Heterocyclic Building Blocks, Pyrrolidines More...
InChI Key   CECTWDBFDAJIEO-NSHDSACASA-N
assay   99%
optical activity   [α]20/D −122°, c = 1 in chloroform
refractive index   n20/D 1.54(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?