• USA Home
  • 348546 - 6-Fluoroveratraldehyde

EMAIL THIS PAGE TO A FRIEND
348546 Aldrich

6-Fluoroveratraldehyde

97%

Purchase

Description

Packaging

1 g in glass bottle

Application

6-Fluoroveratraldehyde may be used in the preparation of 6-[18F]fluoroveratraldehyde via nucleophilic aromatic substitution by [18F]fluoride. It may be used in the preparation of key metabolites of 6-fluoro-DOPA.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?