EMAIL THIS PAGE TO A FRIEND
348716 Aldrich

Silver

foil, thickness 0.25 mm, 99.99% trace metals basis

Purchase

Properties

Related Categories Materials Science, Metal and Ceramic Science, Metals, Silver
InChI Key   BQCADISMDOOEFD-UHFFFAOYSA-N
assay   99.99% trace metals basis
form   foil
resistivity   1.59 μΩ-cm, 20°C
thickness   0.25 mm
bp   2212 °C(lit.)
mp   960 °C(lit.)
density   10.49 g/cm3(lit.)

Description

Packaging

1 ea in rigid mailer

Quantity

50 x 50 mm (approximately 6.6 g)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Ultra-High Purity Inorganics
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?