EMAIL THIS PAGE TO A FRIEND
348767 Aldrich

Silver

rod, diam. 6.35 mm, ≥99.95%

Purchase

Properties

Related Categories Alternative Energy, Materials Science, Metal and Ceramic Science, Metals, Silver,
InChI Key   BQCADISMDOOEFD-UHFFFAOYSA-N
assay   ≥99.95%
form   rod
resistivity   1.59 μΩ-cm, 20°C
diam.   6.35 mm
bp   2212 °C(lit.)
mp   960 °C(lit.)
density   10.49 g/cm3(lit.)

Description

Quantity

17 g = 50 mm; 34 g = 100 mm; 51 g = 150 mm

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Ultra-High Purity Inorganics
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

comparable product

Product #

Description

Add to Cart

265624 Silver, rod, diam. 7.0 mm, 99.99% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?