EMAIL THIS PAGE TO A FRIEND
349429 Aldrich

Zinc

foil, thickness 0.5 mm, 99.999%

Purchase

Properties

Related Categories Materials Science, Metal and Ceramic Science, Metals, Zinc
vapor pressure   1 mmHg ( 487 °C)
InChI Key   HCHKCACWOHOZIP-UHFFFAOYSA-N
assay   99.999%
form   foil
resistivity   5.8 μΩ-cm, 20°C
thickness   0.5 mm
bp   907 °C(lit.)
mp   420 °C(lit.)
density   7.133 g/mL at 25 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

267619 Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?