EMAIL THIS PAGE TO A FRIEND
356875 Aldrich

Cobalt

foil, thickness 0.25 mm, 99.95%

Purchase

Properties

Related Categories 27: Co, Cobalt, Magnetic Materials, Magnetic Precursors, Materials Science,
InChI Key   GUTLYIVDDKVIGB-UHFFFAOYSA-N
assay   99.95%
form   foil
resistivity   6.24 μΩ-cm, 20°C
thickness   0.25 mm
bp   2900 °C(lit.)
density   8.9 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
-
RTECS 
GF8750000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

356891 Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?