CAS Number 7439-88-5
Empirical Formula (Hill Notation) Ir
Molecular Weight 192.22
MDL number MFCD00011062
PubChem Substance ID 24861887
Popular Documents: Specification Sheet (PDF)
| Related Categories | Iridium, Materials Science, Metal and Ceramic Science, Metals |
| assay | 99.9% trace metals basis |
| form | wire |
| resistivity | 4.71 μΩ-cm |
| diam. | 0.5 mm |
| bp | 4130 °C(lit.) |
| mp | 2450 °C(lit.) |
| density | 22.65 g/cm3(lit.) |
130 mg in rigid mailer
130 mg = 30 mm; 650 mg = 150 mm
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?