• USA Home
  • 359564 - 4-Azidophenyl isothiocyanate

EMAIL THIS PAGE TO A FRIEND
359564 Aldrich

4-Azidophenyl isothiocyanate

97%

Purchase

Properties

Related Categories Aromatic Azides, Azides, Building Blocks, Chemical Biology, Chemical Synthesis,
InChI Key   XMPGXADJZXSISQ-UHFFFAOYSA-N
assay   97%
mp   65-68 °C(lit.)
storage temp.   2-8°C

Description

Packaging

500 mg in glass bottle

Application

4-Azidophenyl isothiocyanate may be used in the preparation of:
• peptide conjugated surfaces on silicon surfaces through click chemistry
• kainic acid analog, (2S.3S.4S)-4-[l-(4′-azido-phenyl)thioureylenernethy lethenyl]-2-carboxy-3-pyrro-lidineacetic acid
• photosensitive nanoparticles bearing azidophenyl units at their surface

General description

4-Azidophenyl isothiocyanate is an aromatic azide.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN1325 - class 4.1 - PG 2 - Flammable solids, organic, n.o.s., HI: all
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?