• USA Home
  • 360309 - 1-(4-Nitrophenyl)glycerol

EMAIL THIS PAGE TO A FRIEND
360309 Aldrich

1-(4-Nitrophenyl)glycerol

99%

Synonym: α-p-Nitrophenylglycerol, 1-(4-Nitrophenyl)-1,2,3-propanetriol, PNPG

Purchase

Properties

Related Categories Alcohols, Building Blocks, C9 to C10, Chemical Synthesis, Organic Building Blocks,
InChI Key   IUZVZBIQZKBWCC-UHFFFAOYSA-N
assay   99%
mp   98-101 °C(lit.)

Description

Packaging

50 mg in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

N1143 1-(4-Nitrophenyl)glycerol, crystalline

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?