• USA Home
  • 360341 - Dimethyl pimelimidate dihydrochloride

EMAIL THIS PAGE TO A FRIEND
360341 Aldrich

Dimethyl pimelimidate dihydrochloride

Synonym: DMP, Dimethyl pimelinediimidate dihydrochloride

Purchase

Properties

Related Categories Amine to Amine, Chemical Biology, Chemical Synthesis, Crosslinkers, Linkers and Crosslinkers More...
InChI Key   LRHXBHUTQWIZTN-UHFFFAOYSA-N

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?