• USA Home
  • 361577 - (R)-(−)-Epichlorohydrin

EMAIL THIS PAGE TO A FRIEND
361577 Aldrich

(R)-(−)-Epichlorohydrin

99%

Synonym: (R)-(−)-2-(Chloromethyl)oxirane

  • CAS Number 51594-55-9

  • Empirical Formula (Hill Notation) C3H5ClO

  • Molecular Weight 92.52

  •  Beilstein Registry Number 1420785

  •  EC Number 424-280-2

  •  MDL number MFCD00077759

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

vapor density   3.29 (vs air)
vapor pressure   10 mmHg ( 16.6 °C)
InChI Key   BRLQWZUYTZBJKN-VKHMYHEASA-N
assay   99%
optical activity   [α]20/D −34°, c = 1 in methanol
optical purity   ee: 97% (GLC)
autoignition temp.   772 °F
expl. lim.   21 %
refractive index   n20/D 1.438(lit.)
bp   114 °C(lit.)
density   1.180 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN 2023 3(6.1) / PGII
WGK Germany 
3
RTECS 
RR0427000
Flash Point(F) 
89.6 °F
Flash Point(C) 
32 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replacement part

Product #

Description

Add to Cart

540072 (R)-(−)-Epichlorohydrin, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?