• USA Home
  • 361585 - (S)-(+)-Epichlorohydrin

EMAIL THIS PAGE TO A FRIEND
361585 Aldrich

(S)-(+)-Epichlorohydrin

98%

Synonym: (S)-(+)-2-(Chloromethyl)oxirane

  • CAS Number 67843-74-7

  • Empirical Formula (Hill Notation) C3H5ClO

  • Molecular Weight 92.52

  •  Beilstein Registry Number 1420784

  •  MDL number MFCD00077760

  •  PubChem Substance ID 329755305

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

vapor density   3.2 (vs air)
vapor pressure   12.5 mmHg ( 20 °C)
InChI Key   BRLQWZUYTZBJKN-GSVOUGTGSA-N
assay   98%
optical activity   [α]20/D +34°, c = 1 in methanol
optical purity   ee: 97% (GLC)
autoignition temp.   1421 °F
expl. lim.   21 %
refractive index   n20/D 1.438(lit.)
bp   92-93 °C/360 mmHg(lit.)
density   1.183 g/mL at 25 °C(lit.)

Description

Application

Chiral synthon employed in the preparation of glycidyl ethers.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2023 6.1/PG 2
WGK Germany 
3
RTECS 
RR0427100
Flash Point(F) 
90 °F
Flash Point(C) 
32 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

540080 (S)-(+)-Epichlorohydrin, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?