• USA Home
  • 362638 - Dichloro[(S)-N,N-dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine]palladium(II)

EMAIL THIS PAGE TO A FRIEND
362638 Aldrich

Dichloro[(S)-N,N-dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine]palladium(II)

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Other Chiral Catalysts
InChI Key   CDELYZVUZJXVRD-VCPBINOTSA-L
optical activity   [α]20/D −170°, c = 0.1 in chloroform
activity   ~0.5 mol dichloromethane/mol
mp   170 °C (dec.)(lit.)

Description

Packaging

100 mg in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?