• USA Home
  • 363022 - Methyl-13C phenyl sulfone

EMAIL THIS PAGE TO A FRIEND
363022 Aldrich

Methyl-13C phenyl sulfone

99 atom % 13C

Purchase

Properties

Related Categories Alphabetical Listings, M, Stable Isotopes More...
InChI Key   JCDWETOKTFWTHA-OUBTZVSYSA-N
isotopic purity   99 atom % 13C
mp   85-88 °C(lit.)
mass shift   M+1

Description

Packaging

250 mg in glass bottle

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Application

Stable one-carbon synthon used for preparing labeled ketones.

Price and Availability


Stable Isotope
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
WR6800000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?