EMAIL THIS PAGE TO A FRIEND
363162 Aldrich

Poly(vinyl alcohol)

average Mw 146,000-186,000, 98-99% hydrolyzed

Purchase

Properties

Related Categories Hydrophilic Polymers, Materials Science, Poly(vinyl alcohol) (PVA), Poly(vinyl alcohol) (PVA) and Copolymers, Polymer Science,
mol wt   average Mw 146,000-186,000
falling ball   55-65 cP, 4 % in H2O(20 °C)(lit.)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
TR8100000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?