CAS Number 6130-98-9
Linear Formula (CH3)3CC≡CC≡CC(CH3)3
Molecular Weight 162.27
MDL number MFCD00134265
PubChem Substance ID 24862208
Popular Documents: FTNMR (PDF)
| Related Categories | Alkynes, Building Blocks, Chemical Synthesis, Internal, Organic Building Blocks More... |
| assay | 99% |
| mp | 130-132 °C(lit.) |
1 g in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?