• USA Home
  • 363758 - 4′-Chloro-2-cyanoacetanilide

EMAIL THIS PAGE TO A FRIEND
363758 Aldrich

4′-Chloro-2-cyanoacetanilide

98%

Purchase

Properties

Related Categories Amides, Building Blocks, C8 to C20+, Carbonyl Compounds, Chemical Synthesis,
InChI Key   FLLVVAHFEBGZKD-UHFFFAOYSA-N
assay   98%
mp   207-209 °C(lit.)

Description

Packaging

100 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?