• USA Home
  • 36657 - (+)-N,N-Dicyclohexyl-(1R)-isoborneol-10-sulfonamide

EMAIL THIS PAGE TO A FRIEND
36657 Aldrich

(+)-N,N-Dicyclohexyl-(1R)-isoborneol-10-sulfonamide

≥98.0%

Synonym: (+)-10-Dicyclohexylsulfamoylisoborneol, (1R)-10-(N,N-Dicyclohexylsulfamoyl)isoborneol, (1R)-(+)-Isoborneol-10-sulfonic dicyclohexylamide, 1S-exo-N,N-Dicyclohexyl-2-hydroxy-7,7-dimethylbicyclo[2.2.1]heptane-1-methanesulfonamide

  • CAS Number 96303-88-7

  • Empirical Formula (Hill Notation) C22H39NO3S

  • Molecular Weight 397.61

  •  Beilstein Registry Number 4712057

  •  MDL number MFCD00151335

  •  PubChem Substance ID 24862670

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Sulfonamides/Sulfinamides More...
InChI Key   UOFKHCXOCUOSKA-NQSCKRDGSA-N
assay   ≥98.0%
optical activity   [α]20/D +26±2°, c = 0.8% in ethanol
mp   159-162 °C(lit.)
  159-162 °C

Description

Other Notes

Chiral auxiliary confering excellent diastereodifferentation in reactions of O-enoyl and enolate derivatives

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?