• USA Home
  • 366625 - 5-Fluoro-2-methylbenzoic acid

EMAIL THIS PAGE TO A FRIEND
366625 Aldrich

5-Fluoro-2-methylbenzoic acid

99%

Purchase

Description

Packaging

1 g in glass bottle

Application

5-Fluoro-2-methylbenzoic acid may be used in the preparation of:
• 5-fluoro-3-hydroxy-2-methylbenzoic acid
• 5-fluoro-2-methyl-3-nitrobenzoic acid methyl ester
• 5-fluoro-N,2-dimethylbenzamide

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?