367281 Aldrich

(1S,2R)-1-Methyl cis-1,2,3,6-tetrahydrophthalate

97%

DOWNLOAD MSDS (PDF)

Synonym: (1S,2R)-1-Methyl cis-4-cyclohexene-1,2-dicarboxylate, (1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethylester

  • CAS Number 88335-93-7

  • Linear Formula HO2CC6H8CO2CH3

  • Molecular Weight 184.19

  •  Beilstein Registry Number 5262096

  •  MDL number MFCD00075490

  •  PubChem Substance ID 24862738

  • Popular Documents:  FTNMR (PDF)  

Properties

assay   97%
optical activity   [α]20/D +13°, c = 1 in acetone
mp   65-67 °C(lit.)

Price and Availability

Customers Also Viewed

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?