• USA Home
  • 370975 - Lignosulfonic acid sodium salt

EMAIL THIS PAGE TO A FRIEND
370975 Aldrich

Lignosulfonic acid sodium salt

average Mw 8,000, average Mn 3,000

Purchase

Properties

Related Categories Lignins, Materials Science, Natural Polymers and Biopolymers, Polymer Science, Polymers More...
mol wt   average Mn 3,000
  average Mw 8,000

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?