• USA Home
  • 371327 - 3-Chloro-N,N-bis(trimethylsilyl)aniline

EMAIL THIS PAGE TO A FRIEND
371327 Aldrich

3-Chloro-N,N-bis(trimethylsilyl)aniline

98%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks, Protected Amines More...
InChI Key   QFZHUSXLYYOLGX-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.5(lit.)
bp   85 °C/0.4 mmHg(lit.)
density   0.981 g/mL at 25 °C(lit.)

Description

Packaging

5 g in glass bottle

Application

3-Chloro-N,N-bis(trimethylsilyl)aniline is suitable for the preparation of the corresponding Grignard reagent.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
219.2 °F
Flash Point(C) 
104 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?